2,2-Dipropylpentanoic acid 95%

Supplier: Ambeed
Warning

A337498-1G A337498-250MG
76828-414EA 647 USD
76828-414 76828-416
2,2-Dipropylpentanoic acid 95%
2,2-Dipropylpentanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₂₂O₂
MDL Number: MFCD02258594
CAS Number: 52061-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR