4,5-Bis(2-methoxyethoxy)-2-nitrobenzonitrile 97%

Supplier: Ambeed
Warning

A336481-10G A336481-25G
76828-408EA 24.41 USD
76828-408 76828-410
4,5-Bis(2-methoxyethoxy)-2-nitrobenzonitrile 97%
4,5-Bis(2-methoxyethoxy)-2-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂O₆
MW: 296.27 g/mol
MDL Number: MFCD17676647
CAS Number: 236750-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR