Thieno[2,3-d]pyrimidine 98%

Supplier: Ambeed
Warning

A333898-1G A333898-250MG
76828-384EA 263.11 USD
76828-384 76828-386
Thieno[2,3-d]pyrimidine 98%
Thieno[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₄N₂S
MW: 136.17 g/mol
MDL Number: MFCD11110440
CAS Number: 272-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR