(6-Bromo-1H-indazol-3-yl)methanol 97%

Supplier: Ambeed
Warning

A333533-100MG A333533-250MG
76828-374EA 20.16 USD
76828-374 76828-376
(6-Bromo-1H-indazol-3-yl)methanol 97%
(6-Bromo-1H-indazol-3-yl)methanol

Precaución: for Research Use Only

Formula: C₈H₇BrN₂O
MW: 227.05 g/mol
MDL Number: MFCD07357298
CAS Number: 885518-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR