Methyl benzo[d]thiazole-6-carboxylate 95%

Supplier: Ambeed
Warning

A333403-100MG A333403-1G A333403-250MG
76828-368EA 15 USD
76828-368 76828-370 76828-372
Methyl benzo[d]thiazole-6-carboxylate 95%
Methyl benzo[d]thiazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇NO₂S
MDL Number: MFCD17018863
CAS Number: 73931-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR