7-Methoxy-2H-benzo[b][1,4]oxazin-3(4H)-one 97%

Supplier: Ambeed
Warning

A333309-250MG A333309-5G
76828-364EA 28 USD
76828-364 76828-366
7-Methoxy-2H-benzo[b][1,4]oxazin-3(4H)-one 97%
7-Methoxy-2H-benzo[b][1,4]oxazin-3(4H)-one

Precaución: for Research Use Only

Formula: C₉H₉NO₃
MDL Number: MFCD06252105
CAS Number: 6529-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR