4-Methylquinazoline 95%

Supplier: Ambeed
Warning

A331983-1G A331983-250MG A331983-5G
76828-348EA 242 USD
76828-348 76828-350 77885-873
4-Methylquinazoline 95%
4-Methylquinazoline

Precaución: for Research Use Only

Formula: C₉H₈N₂
MDL Number: MFCD05982001
CAS Number: 700-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR