(E)-Ethyl 3-acetamidobut-2-enoate 95%

Supplier: Ambeed
Warning

A331445-1G A331445-250MG
76828-342EA 690.65 USD
76828-342 76828-344
(E)-Ethyl 3-acetamidobut-2-enoate 95%
(E)-Ethyl 3-acetamidobut-2-enoate

Precaución: for Research Use Only

Formula: C₈H₁₃NO₃
MW: 171.19 g/mol
MDL Number: MFCD02974463
CAS Number: 23652-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR