(4-(Pyridin-2-yl)phenyl)methanamine 95%

Supplier: Ambeed
Warning

A331444-100MG A331444-1G A331444-250MG A331444-5G
76828-334EA 48.81 USD
76828-334 76828-336 76828-338 76828-340
(4-(Pyridin-2-yl)phenyl)methanamine 95%
(4-(Pyridin-2-yl)phenyl)methanamine

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂
MW: 184.23 g/mol
MDL Number: MFCD11932623
CAS Number: 294647-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR