2-(2,5-Difluorophenyl)pyrrolidine 98%

Supplier: Ambeed
Warning

A329086-1G A329086-250MG A329086-5G
76828-320EA 63 USD
76828-320 76828-322 76828-324
2-(2,5-Difluorophenyl)pyrrolidine 98%
2-(2,5-Difluorophenyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₁₀H₁₁F₂N
MDL Number: MFCD05189458
CAS Number: 886503-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR