4-Iodo-2-methoxyphenol 97%

Supplier: Ambeed
Danger

Synonyms: 4-Iodoguaiacol, 2-Hydroxy-5-iodoanisole

A328482-100MG A328482-250MG
76828-316EA 16.98 USD
76828-316 76828-318
4-Iodo-2-methoxyphenol 97%
4-Iodo-2-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇IO₂
MW: 250.034 g/mol
MDL Number: MFCD08061924
CAS Number: 203861-62-5
UN: 2811
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR