2'-Fluoro-4'-hydroxyacetophenone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Fluoro-4-hydroxyphenyl)ethanone

A327783-100G A327783-10G A327783-1G A327783-25G A327783-5G
76828-296EA 235.52 USD
76828-296 76828-298 76828-300 76828-302 76828-304
2'-Fluoro-4'-hydroxyacetophenone 97%
2'-Fluoro-4'-hydroxyacetophenone

Precaución: for Research Use Only

Formula: C₈H₇FO₂
MW: 154.14 g/mol
MDL Number: MFCD00142713
CAS Number: 98619-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR