(11bS)-4-Hydroxy-2,6-bis(triphenylsilyl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 98%

Supplier: Ambeed
Warning

A327179-100MG A327179-1G A327179-250MG A327179-5G
76828-282EA 30 USD
76828-282 76828-284 76828-286 76828-288
(11bS)-4-Hydroxy-2,6-bis(triphenylsilyl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 98%
(11bS)-4-Hydroxy-2,6-bis(triphenylsilyl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

Precaución: for Research Use Only

Formula: C₅₆H₄₁O₄PSi₂
MDL Number: MFCD09265079
CAS Number: 929097-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR