N-Methyl-N-(2-(((2-((2-oxoindolin-5-yl)amino)-5-(trifluoromethyl)pyrimidin-4-yl)amino)methyl)phenyl)methanesulphonamide ≥95%

Supplier: Ambeed
Warning

Synonyms: N-Methyl-N-(2-(((2-((2-oxoindolin-5-yl)amino)-5-(trifluoromethyl)pyrimidin-4-yl)amino)methyl)phenyl)methanesulphonamide

A325541-10MG A325541-1MG A325541-50MG A325541-5MG
76828-256EA 75 USD
76828-256 76828-258 76828-260 76828-262
N-Methyl-N-(2-(((2-((2-oxoindolin-5-yl)amino)-5-(trifluoromethyl)pyrimidin-4-yl)amino)methyl)phenyl)methanesulphonamide ≥95%
PF-431396

Precaución: for Research Use Only

Formula: C₂₂H₂₁F₃N₆O₃S
MW: 506.50 g/mol
MDL Number: MFCD16038300
CAS Number: 717906-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR