Methyl 2-amino-4-(benzyloxy)-5-methoxybenzoate 95%

Supplier: Ambeed
Warning

A325193-100MG A325193-1G A325193-250MG A325193-5G
76828-248EA 25.47 USD
76828-248 76828-250 76828-252 76828-254
Methyl 2-amino-4-(benzyloxy)-5-methoxybenzoate 95%
Methyl 2-amino-4-(benzyloxy)-5-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₇NO₄
MW: 287.31 g/mol
MDL Number: MFCD16620533
CAS Number: 61032-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR