6-[(tert-Butoxycarbonyl)amino]spiro[3.3]heptane-2-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 6-((tert-Butoxycarbonyl)amino)spiro[3.3]heptane-2-carboxylic acid, 6-{[(tert-butoxy)carbonyl]amino}spiro[3.3]heptane-2-carboxylic acid

A323746-1G A323746-250MG A323746-5G A323746-100MG
76828-236EA 153.84 USD
76828-236 76828-238 77612-868 77683-964
6-[(tert-Butoxycarbonyl)amino]spiro[3.3]heptane-2-carboxylic acid 98%
6-[(tert-Butoxycarbonyl)amino]spiro[3.3]heptane-2-carboxylic acid

Precaución: For research use only.

Formula: C₁₃H₂₁NO₄
MW: 255.32 g/mol
MDL Number: MFCD11099888
CAS Number: 1087798-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR