2-(4-Methyl-2-nitrophenyl)acetic acid 95%

Supplier: Ambeed
Warning

A321172-250MG A321172-100MG
76828-212EA 60.48 USD
76828-212 77683-957
2-(4-Methyl-2-nitrophenyl)acetic acid 95%
2-(4-Methyl-2-nitrophenyl)acetic acid

Precaución: For research use only.

Formula: C₉H₉NO₄
MW: 195.17 g/mol
MDL Number: MFCD08276007
CAS Number: 29640-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR