N-Boc-(S)-2-amino-1-butanol 95%

Supplier: Ambeed
Danger

A321044-1G A321044-250MG A321044-25G A321044-5G
76828-188EA 6 USD
76828-188 76828-190 76828-192 77593-902
N-Boc-(S)-2-amino-1-butanol 95%
N-Boc-(S)-2-amino-1-butanol

Precaución: for Research Use Only

Formula: C₉H₁₉NO₃
MDL Number: MFCD04974275
CAS Number: 150736-72-4
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR