6-Bromo-4-chloroquinoline-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A320895-100MG A320895-1G A320895-250MG
76828-182EA 130 USD
76828-182 76828-184 76828-186
6-Bromo-4-chloroquinoline-3-carboxylic acid 95%
6-Bromo-4-chloroquinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₅BrClNO₂
MDL Number: MFCD09746284
CAS Number: 179024-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR