1,2-Dibromo-4-methyl-5-nitrobenzene 95%

Supplier: Ambeed
Warning

A319066-100MG A319066-1G A319066-250MG
76828-158EA 12.74 USD
76828-158 76828-160 76828-162
1,2-Dibromo-4-methyl-5-nitrobenzene 95%
1,2-Dibromo-4-methyl-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅Br₂NO₂
MW: 294.92 g/mol
MDL Number: MFCD23135744
CAS Number: 101580-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR