2-Bromo-5-methylnicotinonitrile 97%

Supplier: Ambeed
Warning

A318959-100MG A318959-250MG
76828-154EA 74.27 USD
76828-154 76828-156
2-Bromo-5-methylnicotinonitrile 97%
2-Bromo-5-methylnicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.03 g/mol
MDL Number: MFCD20925052
CAS Number: 65996-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR