Methyl 5-formylpyrazine-2-carboxylate 98%

Supplier: Ambeed
Warning

A318228-100MG A318228-1G A318228-250MG A318228-5G
76828-144EA 10 USD
76828-144 76828-146 76828-148 76828-150
Methyl 5-formylpyrazine-2-carboxylate 98%
Methyl 5-formylpyrazine-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₆N₂O₃
MW: 166.134 g/mol
MDL Number: MFCD12755910
CAS Number: 710322-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR