(2,4-Difluorophenyl)-4-piperidylmethanone oxime hydrochloride 95%

Supplier: Ambeed
Danger

Synonyms: (2,4-Difluorophenyl)(piperidin-4-yl)methanone oxime hydrochloride

A318166-10G A318166-25G A318166-5G
76828-132EA 20.16 USD
76828-132 76828-134 76828-136
(2,4-Difluorophenyl)-4-piperidylmethanone oxime hydrochloride 95%
(2,4-Difluorophenyl)-4-piperidylmethanone oxime hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₄F₂N₂O
MW: 276.71 g/mol
MDL Number: MFCD11869788
CAS Number: 135634-18-3
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR