3-Bromo-2-chloro-5-thiophenesulphonyl chloride 97%

Supplier: Ambeed
Danger

Synonyms: 4-Bromo-5-chlorothiophene-2-sulphonyl chloride

A317581-5G A317581-250MG
76828-116EA 155 USD
76828-116 77885-848
3-Bromo-2-chloro-5-thiophenesulphonyl chloride 97%
3-Bromo-2-chloro-5-thiophenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₄HBrCl₂O₂S₂
MW: 295.9895 g/mol
MDL Number: MFCD00052094
CAS Number: 166964-35-8
UN: 3261
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR