4-(Methylcarbamoyl)benzene-1-sulfonyl chloride 95%

Supplier: Ambeed
Danger

A316486-100MG A316486-1G A316486-250MG
76828-092EA 37.14 USD
76828-092 76828-094 76828-096
4-(Methylcarbamoyl)benzene-1-sulfonyl chloride 95%
4-(Methylcarbamoyl)benzene-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₈H₈ClNO₃S
MW: 233.67 g/mol
MDL Number: MFCD07691322
CAS Number: 874622-79-4
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR