6-Chloro-5-(trifluoromethyl)nicotinaldehyde 95%

Supplier: Ambeed
Warning

A316411-100MG A316411-1G A316411-5G
76828-086EA 171.87 USD
76828-086 76828-088 76828-090
6-Chloro-5-(trifluoromethyl)nicotinaldehyde 95%
6-Chloro-5-(trifluoromethyl)nicotinaldehyde

Precaución: for Research Use Only

Formula: C₇H₃ClF₃NO
MW: 209.55 g/mol
MDL Number: MFCD18258310
CAS Number: 1113049-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR