(R)-3-Amino-2-benzylpropanoic acid 98%

Supplier: Ambeed
Warning

A316357-100MG A316357-1G A316357-250MG
76828-080EA 106.09 USD
76828-080 76828-082 76828-084
(R)-3-Amino-2-benzylpropanoic acid 98%
(R)-3-Amino-2-benzylpropanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
MDL Number: MFCD07372494
CAS Number: 183182-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR