S-Methyl-L-cysteine 98%

Supplier: Ambeed
Warning

Synonyms: H-Cys(Me)-OH

A316051-1G A316051-250MG A316051-25G A316051-5G
76828-072EA 16 USD
76828-072 76828-074 76828-076 76828-078
S-Methyl-L-cysteine 98%
S-Methyl-L-cysteine

Precaución: for Research Use Only

Formula: C₄H₉NO₂S
MDL Number: MFCD00002612
CAS Number: 1187-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR