Ethyl 3-azabicyclo[3.1.0]hexane-6-carboxylate hydrochloride 95%

Supplier: Ambeed
Warning

A315691-1G A315691-250MG
76828-068EA 476.35 USD
76828-068 76828-070
Ethyl 3-azabicyclo[3.1.0]hexane-6-carboxylate hydrochloride 95%
Ethyl 3-azabicyclo[3.1.0]hexane-6-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₃NO₂·ClH
MW: 191.655 g/mol
MDL Number: MFCD11111227
CAS Number: 1211510-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR