(3R,5S)-rel-3,5-Dimethylpiperidine 98%

Supplier: Ambeed
Danger

A314995-1G A314995-25G A314995-250MG A314995-5G
76828-064EA 144.29 USD
76828-064 76828-066 77683-935 77683-936
(3R,5S)-rel-3,5-Dimethylpiperidine 98%
(3R,5S)-rel-3,5-Dimethylpiperidine

Precaución: For research use only.

Formula: C₇H₁₅N
MW: 113.20 g/mol
MDL Number: MFCD09832871
CAS Number: 14446-75-4
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR