(S)-Pyrrolidine-2-carboxylic acid hydrochloride 98%

Supplier: Ambeed
Warning

A313324-1G A313324-25G A313324-5G
76828-046EA 21.22 USD
76828-046 76828-048 76828-050
(S)-Pyrrolidine-2-carboxylic acid hydrochloride 98%
(S)-Pyrrolidine-2-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₀ClNO₂
MW: 151.59 g/mol
MDL Number: MFCD03791121
CAS Number: 7776-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR