3-Amino-2-methylphenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

A312916-100MG A312916-1G A312916-250MG A312916-5G
76828-030EA 23.34 USD
76828-030 76828-032 76828-034 76828-036
3-Amino-2-methylphenylboronic acid pinacol ester 98%
3-Amino-2-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₀BNO₂
MW: 233.12 g/mol
MDL Number: MFCD11054038
CAS Number: 882678-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR