1-(2-Bromo-5-methylphenyl)ethan-1-one 95%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Bromo-5-methylphenyl)ethanone

A312053-100MG A312053-1G A312053-250MG
76828-022EA 59.42 USD
76828-022 76828-024 76828-026
1-(2-Bromo-5-methylphenyl)ethan-1-one 95%
1-(2-Bromo-5-methylphenyl)ethan-1-one

Precaución: for Research Use Only

Formula: C₉H₉BrO
MW: 213.071 g/mol
MDL Number: MFCD11695376
CAS Number: 77344-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR