2-(4-Bromophenyl)benzimidazole 97%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Bromophenyl)-1H-benzo[d]imidazole

A311468-1G A311468-5G
76828-004EA 83.82 USD
76828-004 76828-006
2-(4-Bromophenyl)benzimidazole 97%
2-(4-Bromophenyl)benzimidazole

Precaución: for Research Use Only

Formula: C₁₃H₉BrN₂
MW: 273.13 g/mol
MDL Number: MFCD00224360
CAS Number: 2622-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR