Methyl-8-chloro-4-oxo-1,4-dihydroquinoline-7-carboxylate 96%

Supplier: Ambeed
Warning

Synonyms: Methyl 8-chloro-4-oxo-1,4-dihydroquinoline-7-carboxylate

A310802-100MG A310802-250MG
76827-994EA 38.2 USD
76827-994 76827-996
Methyl-8-chloro-4-oxo-1,4-dihydroquinoline-7-carboxylate 96%
Methyl-8-chloro-4-oxo-1,4-dihydroquinoline-7-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₈ClNO₃
MW: 237.6400 g/mol
MDL Number: MFCD11501427
CAS Number: 948573-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR