(2,6-Dimethylpyridin-3-yl)methanol 97%

Supplier: Ambeed
Warning

A310042-1G A310042-250MG
76827-986EA 33 USD
76827-986 76827-988
(2,6-Dimethylpyridin-3-yl)methanol 97%
(2,6-Dimethylpyridin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MDL Number: MFCD03265779
CAS Number: 582303-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR