Methyl 4-(benzyloxy)-5-methoxy-2-nitrobenzoate 95%

Supplier: Ambeed
Warning

A309910-100G A309910-10G A309910-1G A309910-250MG A309910-25G A309910-5G
76827-974EA 241.89 USD
76827-974 76827-976 76827-978 76827-980 76827-982 76827-984
Methyl 4-(benzyloxy)-5-methoxy-2-nitrobenzoate 95%
Methyl 4-(benzyloxy)-5-methoxy-2-nitrobenzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₅NO₆
MW: 317.29 g/mol
MDL Number: MFCD18207027
CAS Number: 61032-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR