6-Amino-5-nitronicotinic acid 98%

Supplier: Ambeed
Warning

A308249-10G A308249-1G A308249-5G A308249-250MG
76827-960EA 235 USD
76827-960 76827-962 76827-964 77885-820
6-Amino-5-nitronicotinic acid 98%
6-Amino-5-nitronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₅N₃O₄
MDL Number: MFCD11100828
CAS Number: 89488-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR