6-Chloro-5-methylpyrimidine-2,4(1H,3H)-dione 97%

Supplier: Ambeed
Warning

A307819-100MG A307819-1G A307819-250MG
76827-954EA 49.87 USD
76827-954 76827-956 76827-958
6-Chloro-5-methylpyrimidine-2,4(1H,3H)-dione 97%
6-Chloro-5-methylpyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O₂
MW: 160.56 g/mol
MDL Number: MFCD01011765
CAS Number: 1627-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR