1-(3,4-Dimethylphenyl)-3-methyl-5-pyrazolone 95%

Supplier: Ambeed
Warning

Synonyms: 2-(3,4-Dimethylphenyl)-5-methyl-1H-pyrazol-3(2H)-one

A306666-1G A306666-5G
76827-946EA 18.04 USD
76827-946 76827-948
1-(3,4-Dimethylphenyl)-3-methyl-5-pyrazolone 95%
1-(3,4-Dimethylphenyl)-3-methyl-5-pyrazolone

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O
MW: 202.26 g/mol
MDL Number: MFCD09795980
CAS Number: 277299-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR