(3-((2-Chlorophenoxy)methyl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A306441-1G A306441-250MG A306441-5G
76827-940EA 137.92 USD
76827-940 76827-942 76827-944
(3-((2-Chlorophenoxy)methyl)phenyl)boronic acid 97%
(3-((2-Chlorophenoxy)methyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂BClO₃
MW: 262.50 g/mol
MDL Number: MFCD16198497
CAS Number: 1256358-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR