Ethyl-N-methyl-β-alaninate 95%

Supplier: Ambeed
Warning

A304352-10G A304352-1G A304352-5G A304352-250MG
76827-924EA 101 USD
76827-924 76827-926 76827-928 77612-752
Ethyl-N-methyl-β-alaninate 95%
Ethyl-N-methyl-beta-alaninate

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂
MDL Number: MFCD00672194
CAS Number: 2213-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR