(R)-4-tert-Butyl 3-methyl morpholine-3,4-dicarboxylate 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A303310-100MG A303310-250MG
76827-910EA 59.42 USD
76827-910 77683-903
(R)-4-tert-Butyl 3-methyl morpholine-3,4-dicarboxylate 97%, Ambeed.Inc
(R)-4-tert-Butyl 3-methyl morpholine-3,4-dicarboxylate

Precaución: For research use only.

Formula: C₁₁H₁₉NO₅
MW: 245.27 g/mol
MDL Number: MFCD09953091
CAS Number: 885321-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR