(S)-1,1,1-Trifluoro-N-(pyrrolidin-2-ylmethyl)methanesulfonamide 97%

Supplier: Ambeed
Danger

A303117-100MG A303117-1G A303117-250MG A303117-5G
76827-904EA 12 USD
76827-904 76827-906 76827-908 77885-807
(S)-1,1,1-Trifluoro-N-(pyrrolidin-2-ylmethyl)methanesulfonamide 97%
(S)-1,1,1-Trifluoro-N-(pyrrolidin-2-ylmethyl)methanesulfonamide

Precaución: for Research Use Only

Formula: C₆H₁₁F₃N₂O₂S
MDL Number: MFCD11656045
CAS Number: 782495-18-5
UN: 3263
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR