2-(Ethoxycarbonyl)-4-fluorophenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (2-(Ethoxycarbonyl)-4-fluorophenyl)boronic acid

A301204-1G A301204-250MG A301204-5G
76827-866EA 72 USD
76827-866 76827-868 76827-870
2-(Ethoxycarbonyl)-4-fluorophenylboronic acid 97%
2-(Ethoxycarbonyl)-4-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BFO₄
MDL Number: MFCD11053864
CAS Number: 1072951-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR