2-(Dimethylamino)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinonitrile 96%

Supplier: Ambeed
Warning

A300593-100MG A300593-1G A300593-250MG A300593-5G
76827-858EA 37.14 USD
76827-858 76827-860 76827-862 76827-864
2-(Dimethylamino)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinonitrile 96%
2-(Dimethylamino)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinonitrile

Precaución: for Research Use Only

Formula: C₁₄H₂₀BN₃O₂
MW: 273.14 g/mol
MDL Number: MFCD16995282
CAS Number: 1346809-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR