1-((2R,3R,4R,5R)-4-Hydroxy-5-(hydroxymethyl)-3-(2-methoxyethoxy)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione 95%

Supplier: Ambeed
Warning

Synonyms: 2'-O-(2-Methoxyethyl)-uridine

A299991-100MG A299991-1G A299991-250MG A299991-5G
76827-840EA 20.16 USD
76827-840 76827-842 76827-844 77550-822
1-((2R,3R,4R,5R)-4-Hydroxy-5-(hydroxymethyl)-3-(2-methoxyethoxy)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione 95%
1-((2R,3R,4R,5R)-4-Hydroxy-5-(hydroxymethyl)-3-(2-methoxyethoxy)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O₇
MW: 302.28 g/mol
MDL Number: MFCD17215636
CAS Number: 223777-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR