MK-4827 98%

Supplier: Ambeed
Danger

Synonyms: (S)-2-(4-(Piperidin-3-yl)phenyl)-2H-indazole-7-carboxamide, Niraparib

A298513-100MG A298513-10MG A298513-1G A298513-1MG A298513-250MG A298513-2MG A298513-5G A298513-5MG
76827-818EA 46.68 USD
76827-818 76827-820 76827-822 76827-824 76827-826 76827-828 76827-830 76827-832
MK-4827 98%
MK-4827

Precaución: for Research Use Only

Formula: C₁₉H₂₀N₄O
MW: 320.39 g/mol
MDL Number: MFCD17779309
CAS Number: 1038915-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR