6,6-Dimethyl-3-azabicyclo[3.1.0]hexane-2,4-dione 97%

Supplier: Ambeed
Warning

A298008-100MG A298008-1G A298008-250MG A298008-25G
76827-808EA 6.37 USD
76827-808 76827-810 76827-812 76827-814
6,6-Dimethyl-3-azabicyclo[3.1.0]hexane-2,4-dione 97%
6,6-Dimethyl-3-azabicyclo[3.1.0]hexane-2,4-dione

Precaución: for Research Use Only

Formula: C₇H₉NO₂
MDL Number: MFCD12923551
CAS Number: 194421-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR