2',4,4',5',7,7'-Hexachloro-3',6'-dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-6-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 6-HEX

A297995-250MG A297995-1G A297995-5G
76827-806EA 144.29 USD
76827-806 77612-716 77683-888
2',4,4',5',7,7'-Hexachloro-3',6'-dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-6-carboxylic acid 95%
2',4,4',5',7,7'-Hexachloro-3',6'-dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-6-carboxylic acid

Precaución: For research use only.

Formula: C₂₁H₆Cl₆O₇
MW: 582.98 g/mol
MDL Number: MFCD08459569
CAS Number: 155911-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR